Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picloram, Restek
SDP

Catalog No. 06719885 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-885 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-885 Supplier Restek Corporation Supplier No. 32265
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2-1-1918
Molecular Formula C6H3Cl3N2O2
Molecular Weight (g/mol) 241.45
InChI Key NQQVFXUMIDALNH-UHFFFAOYSA-N
PubChem CID 15965
ChEBI CHEBI:34922
IUPAC Name 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
SMILES C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N

Specifications

Quantity 1 mL
Chemical Name or Material Picloram
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.