Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picral Etch 4% Picric Acid in Ethanol, Reagents

Picral Etch 4% Picric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CP122100500C

Quantity 500 mL

 View more versions of this product

Catalog No. 01-334-279


Add to Cart

Description

Description

Specifications

Chemical Identifiers

64-17-5 , 67-63-0 , 67-56-1
46.07
2,4,6-Trinitrophenol
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol

Specifications

Yellow
76°C
2,4,6-Trinitrophenol
CCO
46.07
Reagent
0.8123
500 mL
Liquid
64-17-5 , 67-63-0 , 67-56-1
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
4%
Glass Bottle
1.3
Non-sterile
Picral Etch 4% Picric Acid in Ethanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.