Learn More
Picral Etch 4% Picric Acid in Ethanol, Reagents
Picral Etch 4% Picric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$212.50 - $300.00
Chemical Identifiers
| CAS | 64-17-5 , 67-63-0 , 67-56-1 |
|---|---|
| Molecular Formula | C2H6O |
| Molecular Weight (g/mol) | 46.07 |
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| Synonym | 2,4,6-Trinitrophenol |
| IUPAC Name | ethanol |
| SMILES | CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-334-280
|
Reagents Holdings Llc
CP1221001C |
1 L |
Each for $300.00
|
|
|||||
|
01-334-279
|
Reagents Holdings Llc
CP122100500C |
500 mL |
Each for $212.50
|
|
|||||
Chemical Identifiers
| C2H6O | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| ethanol |
Specifications
| Yellow | |
| 76°C | |
| 2,4,6-Trinitrophenol | |
| CCO | |
| 46.07 | |
| Reagent | |
| 0.8123 | |
| Non-sterile | |
| Picral Etch 4% Picric Acid in Ethanol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.