Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picral Etch 4% Picric Acid in Ethanol, Reagents

Catalog No. 01334280
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-280 1 L
01-334-279 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-280 Supplier Reagents Holdings Llc Supplier No. CP1221001C
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Picral Etch 4% Picric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym 2,4,6-Trinitrophenol
IUPAC Name ethanol
SMILES CCO

Specifications

Color Yellow
Boiling Point 76°C
Percent Purity 4%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8123
pH 1.3
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Picral Etch 4% Picric Acid in Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.