Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picric Acid, Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCP0300125 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 g
25 g
125 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCP0300125 125 g
RDCP030010C 10 g
RDCP030025C 25 g
RDCP0300500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RDCP0300125 Supplier Ricca Chemical Company Supplier No. RDCP0300125A6
Add to Cart
Add to Cart

For safety in transportation, Picric Acid is usually mixed with a minimum of 30% water.

Chemical Identifiers

CAS 7732-18-5 , 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) Mixture
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
IUPAC Name 2,4,6-trinitrophenol

Specifications

CAS Max % 70
SMILES OC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 49%
Grade Reagent
Quantity 125 g
Packaging Natural Poly Bottle
Chemical Name or Material Picric Acid
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.