Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picric Acid, Saturated (AOAC), Ricca Chemical
SDP

Catalog No. SP9200100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP9200-100 100 mL Natural Poly Bottle
SP920050 50 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SP9200-100 Supplier Ricca Chemical Company Supplier No. SP9200100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5 , 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) Mixture
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
Synonym picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
PubChem CID 6954
ChEBI CHEBI:46149
IUPAC Name 2,4,6-trinitrophenol
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Min % 97.0, 1.26
CAS Max % 100.0, 1.31
Grade Laboratory
Quantity 100 mL
Packaging Natural Poly Bottle
Chemical Name or Material Picric Acid, Saturated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.