Learn More
Selleck Chemical LLC Pictilisib (GDC-0941) 10mM/1mL 957054-30-7 RG7321
SDP
Supplier:  Selleck Chemical LLC S106510MM/1ML
Encompass_Preferred
Pictilisib (GDC-0941, RG7321) is a potent inhibitor of PI3K?/? with IC50 of 3 nM in cell-free assays, with modest selectivity against p110? (11-fold) and p110? (25-fold). Pictilisib (GDC-0941) induces autophagy and apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.