Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLL Antibody [10F8D7] , ProSci
SDP

Catalog No. 16835836
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-835-836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-835-836 Supplier ProSci LLC Supplier No. 321940100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLL Monoclonal Antibody specifically detects SORL1 in Human Samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MLL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 10F8D7
Conjugate Unconjugated
Dilution Western Blot:1:500 - 1:2,000 , IHC(P):1:500 - 1:2,000 , ELISA:Propose dilution 1:10,000.
Formulation Ascitic fluid containing 0.03% sodium azide.
Gene Accession No. Q03164
Gene Alias ALL1, CXXC7, HRX, HTRX, MLL, MLL1, TRX1, ALL-1, Lysine N-methyltransferase 2A, HTRX1, MLL1A, WDSTS, MLL/GAS7, TET1-MLL
Host Species Mouse
Immunogen Purified truncated recombinant MLL-Trx (aa3751-3968) is expressed in E.Coli strain BL21 (DE3).
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4297
Target Species Human
Content And Storage MLL monoclonal antibody can be stored at -20°C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Purified
Isotype IgG1
Research Category Other
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.