Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Picus™ NxT Electronic Pipettes, Single Channel

Catalog No. 14557821 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
0.2 to 10 μL
10 to 300 μL
50 to 1000 μL
Color:
Blue
Gray
Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
14-557-821 0.2 to 10 μL Gray
14-557-823 10 to 300 μL Orange
14-557-824 50 to 1000 μL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-557-821 Supplier Sartorius Supplier No. LH745021

Picus™ NxT Electronic Pipettes, Single Channel are designed to ease user's workload and to protect from repetitive strain injury (RSI).

Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.

  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese

  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management

Applications:

PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture

Certifications

CE Marked

Warranty and Services

2 Year

TRUSTED_SUSTAINABILITY

Specifications

Calibration Points Single
Certifications/Compliance Certificate of accredited 3-point calibration
Compatible Tips Universal Design, Safetyspace, Optifit
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
Number of Channels 1
Product Line Picus NxT
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 0.2 to 10 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.