Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 1.1 mL Microtube System, Racked

Catalog No. PI15082 Shop All Thermo Scientific Products
Change view
Click to view available options
Tubes racked in boxes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PI15082 Tubes racked in boxes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15082 Supplier Thermo Scientific™ Supplier No. 15082

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Pre-racked 1.1 mL-tall microtubes boxed to match a standard 96-well microplate footprint for multichannel pipetting or automated workstations.

Thermo Scientific™ 1.1mL Microtubes and Racked Systems consist of 1.1mL tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.

The microtubes resembles 1mL pipette tips except that they have round bottoms. Like a box of 1mL pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.

Highlights:

  • 8 x 12 format simplifies and speeds applications in which a multi-channel pipettor is used
  • Format is also compatible with robotic workstations
  • Thermo Scientific Microtube System can be centrifuged
TRUSTED_SUSTAINABILITY

Specifications

Product Line Thermo Scientific
Quantity 10 boxes
Product Type Microtube Racked System
Description Pierce 1.1mL Microtube Racked System
Sterility Non-sterile
Content And Storage Store at room temperature.
Volume (Metric) 1.1 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.