Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Agarose I (Molecular Biology Grade)

Catalog No. p-4530638
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17850 100 g
PI17852 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI17850 Supplier Thermo Scientific™ Supplier No. 17850
Add to Cart
Add to Cart

Ensure uniform lot-to-lot consistency in preparing electrophoresis gels using highly purified and blended regular- or low-melting agarose formulations.

Thermo Scientific™ Pierce Agaroses are highly purified and carefully blended formulations of regular- or low-melting agarose that provide uniform lot-to-lot consistency for preparation of electrophoresis gels.

Highlights:

  • Agarose I (Molecular Biology Grade)
  • A regular melting-temperature agarose for electrophoresis requiring an exceptional level of purity with unequaled performance
  • Agarose II (Low Melt)
  • A low temperature agarose for special applications requiring a lower melting temperature
  • Optimal for “in-gel” restriction digestion/ligation and DNA recovery
  • At 1%, forms a very manageable clear gel that provides excellent resolution and melting characteristics
  • Melts at approximately 65°C and remains fluid at 37°C for at least 24 hours, providing an excellent matrix for enzymatic manipulation of samples
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Gel Electrophoresis
Melting Point Standard Melting Point
Product Type Agarose I (Molecular Biology Grade)
Quantity 100 g
Separation Range 100 bp to >30 kb

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.