Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Miser™ Antibody Extender Solution NC

Catalog No. PI32110 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
ELISA, Western Blot
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
PI32110 ELISA, Western Blot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI32110 Supplier Thermo Scientific™ Supplier No. 32110
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Enable Western blot detection with much less primary antibody by treating nitrocellulose membrane with this reagent.

Perform Western blot detection with much less primary antibody using Thermo Scientific™ Pierce Antibody Extender Solution NC, a post-transfer nitrocellulose membrane-treatment reagent.

By treating nitrocellulose membranes for a few minutes in this reagent immediately after transfer and before probing, you can use much less (more dilute) primary antibody solutions while obtaining the same level of detection. This conserves use of expensive and rare antibodies, enabling more experiments to be performed with the same amount of antibody. The reagent is so effective that our scientists have observed antibody cost reductions of 10-, 25- and even 100-times (depending on the antigen and primary antibody directed against that antigen).

Highlights:

  • Achieve equivalent signal – use at least three times less primary antibody than normal and retain comparable or better signal detectability

  • Any detection mode or enzyme – the method works with both colorimetric and chemiluminescent detection modes and with horseradish peroxidase (HRP) and alkaline phosphatase (AP) systems

  • Easy-to-perform, 10-minute protocol – invest a few minutes and save money on your primary antibody; our scientists have reduced their use of expensive primary antibodies by up to 100-fold with this product

TRUSTED_SUSTAINABILITY

Specifications

Product Line Miser
Membrane Nitrocellulose Membrane
Form Liquid
Product Type Antibody Extender Solution
Content And Storage Store at room temperature.
For Use With (Application) ELISA, Western Blot
Which should I use: the Pierce Western Blot Signal Enhancer or the Miser Antibody Extender Solution NC?

If you are happy with your western signal, you can use the Miser Antibody Extender Solution NC to minimize the amount of antibody you need to use and therefore reduce your costs. If you need to increase sensitivity, then you can use the Pierce Western Blot Signal Enhancer.

How does the Miser Antibody Extender Solution NC allow me to use less antibody?

The proteins on the blot become more accessible to the antibodies, making it possible for a signal to be produced using less antibody.

Can I use Ponceau S staining before using the Miser Antibody Extender Solution NC?

The Extender Solution should be used after staining with Ponceau S.

Can I use the Miser Antibody Extender Solution NC on nitrocellulose and PVDF membranes?

The Extender solution gives consistent, reproducible results on nitrocellulose membranes; results can vary with PVDF membrane.

When in my workflow do I need to used the Miser Antibody Extender Solution NC?

The Extender Solution should be used after transferring the protein to the membrane and before blocking the membrane.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.