Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BS(PEG)9

Catalog No. PI21582 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21582 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21582 Supplier Thermo Scientific™ Supplier No. 21582
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Crosslink amines to amines with these long (21.7A and 35.8A) crosslinkers composed of NHS-ester groups and 5-unit or 9-unit PEG spacer arms that enhance protein solubility.

Thermo Scientific™ Pierce BS(PEG)n Crosslinkers include BS(PEG)5 and BS(PEG)9, which are bis-succinimide ester-activated PEG compounds for crosslinking between primary amines in proteins and other molecules.

BS(PEG)9 is a bis-succinimide ester-activated PEG compound used for crosslinking between primary amines (–NH2) in proteins and other molecules. The N-hydroxysuccinimide ester (NHS) groups at either end of the PEG9 spacer react specifically and efficiently with lysine and N-terminal amino groups at pH 7-9 to form stable amide bonds.

Features of BS(PEG)9:

  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • NHS esters react with –NH2 groups at pH 7-9, forming stable amide bonds
  • Water-soluble; compare to non-pegylated BS3 (also soluble) (Cat. No. 21580) and DSS (water-insoluble) (Cat. No. 21655)
  • Membrane-impermeable, allowing for cell surface labeling
  • Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
  • Polyethylene glycol spacer arms help maintain conjugate solubility
  • Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Ideal for small molecule or peptide conjugations

Alternative Names for BS(PEG)n:

  • Bis-NHS-dPEG
  • NHS-PEG-NHS
  • BS(PEO)5
  • Bis(NHS)PEO5
  • BS-PEO9
  • dPEG NHS ester
  • Bis-succinimide ester PEG 400

Product References

  • Crosslinker Application Guide -- search for recent literature references for this product
  • Bioconjugate Techniques, Academic Press (Cat. No. 20036)

Related Products

  • BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate) (Cat. No. 21581)
  • BS3 (bis(sulfosuccinimidyl)suberate) (Cat. No. 21580)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description BS(PEG)9
Molecular Weight (g/mol) 708.71
PEGylated Yes
Spacer Arm Length 35.8 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Long (>30 Å)
Form Liquid
Quantity 100 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.