Learn More
Thermo Scientific™ BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce BS(PEG)n Crosslinkers include BS(PEG)5 and BS(PEG)9, which are bis-succinimide ester-activated PEG compounds for crosslinking between primary amines in proteins and other molecules.
BS(PEG)5 is a bis-succinimide ester-activated PEG compound used for crosslinking between primary amines (—NH2) in proteins and other molecules. The N-hydroxysuccinimide ester (NHS) groups at either end of the PEG5 spacer react specifically and efficiently with lysine and N-terminal amino groups at pH 7-9 to form stable amide bonds.
Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.
Features of BS(PEG)5:
- Reactive groups: NHS ester (both ends)
- Reactive towards: amino groups (primary amines)
- NHS esters react with—NH2 groups at pH 7-9, forming stable amide bonds
- Water-soluble; compare to non-pegylated BS3 (also soluble) (Cat. No. 21580) and DSS (water-insoluble) (Cat. No. 21655)
- Membrane-impermeable, allowing for cell surface labeling
- Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
- Polyethylene glycol spacer arms help maintain conjugate solubility
- Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Ideal for small molecule or peptide conjugations
Alternative Names for BS(PEG)n:
- Bis-NHS-dPEG
- NHS-PEG-NHS
- BS(PEO)5
- Bis(NHS)PEO5
- BS-PEO9
- dPEG NHS ester
- Bis-succinimide ester PEG 400
Product References
- Crosslinker Application Guide -- search for recent literature references for this product
- Bioconjugate Techniques, Academic Press (Cat. No. 20036)
Related Products
- BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate) (Cat. No. 21582)
- BS3 (bis(sulfosuccinimidyl)suberate) (Cat. No. 21580)
Specifications
Specifications
| Chemical Reactivity | Amine-Amine |
| Cleavable | No |
| Description | BS(PEG)5 |
| Molecular Weight (g/mol) | 532.5 |
| PEGylated | Yes |
| Spacer Arm Length | 21.7 Å |
| Content And Storage | Upon receipt store desiccated at -20°C. |
| Cell Permeability | No |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
Frequently Asked Questions (FAQs)
BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Amine-to-amine crosslinkers are a good choice for your application since amino groups are usually abundant on the surface of proteins. Our most popular amine-reactive homobifunctional crosslinker that is water-soluble is BS3 (Cat. No. A39266). This has a moderate-length (11.4 A) linker that is good for linking adjacent proteins. If you need the ability to cleave the linker with sulfhydryl reducing agents I would suggest DTSSP (Cat. No. 21578). If you think you might need an extra-long crosslinker, then BS(PEG)5 (Cat. No. 21581) or BS(PEG)9 (Cat. No. 21582) are alternatives.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.