Learn More
Thermo Scientific™ Pierce™ DiscoverLight™ Protein Array Kits
Catalog No. PI81000
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
PI81000 1 Kit
1 options
Catalog No. PI81000 Supplier Thermo Scientific™ Supplier No. 81000
Includes
Includes:
  • DiscoverLight Nitrocellulose Membrane, 4 each
  • 10X Blocker™ BSA in PBS, 50mL
  • Probe (see table)
  • BupH™ Phosphate Buffered Saline, 8 packs
  • 10% Tween™-20, 2 x 10mL
  • SuperSignal West Pico Luminol Enhancer, 2 x 25mL
  • SuperSignal West Pico Stable Peroxide, 2 x 25mL
Convenient, easy-to-use kits for arraying proteins in a low-density format
Utilize specially designed nitrocellulose membranes with a grided template based on a 384-well plate. Eighteen positive control elements consisting of biotin (in yellow) and peroxidase (in blue) serve as positional or functional markers throughout the membrane sheet. The sheet is designed so that it may be segmented for smaller or multiple screens.
  • Screen many samples with minimal probe, buffer, and detection reagent
  • Can be processed using common lab equipment
  • Use SuperSignal™ West Pico Chemiluminescent Substrate† for picogram-level sensitivity and signal within five minutes
  • Can be processed in four hours
  • Ideal for protein profiling, protein expression level monitoring, protein detection, post-translational modification screens, and protein interaction investigation
†SuperSignal Technology is protected by U.S. Patent 6,432,662.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit