Learn More
Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format
Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.
Manufacturer:  Thermo Scientific™ 21655
 View more versions of this product
Catalog No. PI21655
Add to cart
Description
Description
Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.
Highlights:
  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Crosslink oriented antibodies to Protein A or G agarose
  • Immobilize proteins onto amine-coated surfaces
Specifications
Specifications
Pierce™
Homobifunctional
Mid-Length
50 mg
DMF, DMSO
Chemical Labeling
NHS Ester
Powder
Crosslinker