Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Multispecies ERK1/ERK2 In-Cell ELISA Kit, Near Infrared

Catalog No. PI62211
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI62211 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI62211 Supplier Invitrogen™ Supplier No. 62211

ELISA

The ERK1/2 Multispecies In-Cell ELISA Near Infrared Detection Kits provide a simple and convenient method for quantifying intracellular proteins in whole cells. Principle of the method This kit enables simultaneous detection of post-translational-modified protein (PTM) and the corresponding unmodified protein, or two different targets within the same well. Simultaneously detecting targets within an ELISA well eliminates variability caused by differences in cell plating. The expression levels of the protein(s) are monitored using primary antibodies specific to the targets and corresponding species-specific near-infrared Thermo Scientific DyLight-Conjugated Secondary Antibodies. Relative protein activation or inactivation is determined as a ratio of modified protein levels to the corresponding unmodified protein measured using a modification-specific antibody and an antibody to unmodified protein, respectively. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27361, P28482
Description Pierce ERK1/2 Near Infrared In-Cell ELISA Kit
Conjugate DyLight™ 680, DyLight™ 800
Immunoassay Kit Format Multi-component kit with fluorescent conjugates
Product Type ELISA
Sample Type Supernatant
Sufficient For 96 microplate wells of overnight-cultured cells
Target Species Human
For Use With (Equipment) Fluorescent Microplate Reader
Gene ID (Entrez) 5594, 5595
Gene Symbol ERK,ERK-2,ERK2,ERT1,MAPK2,P42MAPK,PRKM1,PRKM2,p38,p40,p41,p41mapk,p42-MAPK,ERK-1,ERK1,ERT2,HS44KDAP,HUMKER1A,P44ERK1,P44MAPK,PRKM3,p44-ERK1,p44-MAPK
Kit Contents Blocking Buffer, 20 x Tris Buffered Saline, Surfact-Amps 20, Surfact-Amps X-100, DyLight® 680 Goat Anti-Mouse IgG (H+L), DyLight 800 Goat Anti-Rabbit IgG (H+L), Thin Plate Seal Assembly, Components included only in the target-specific kits: Antibody #1 and Antibody #2
Label or Dye DyLight™ 680, DyLight™ 800
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Supernatant 10,000 cells
Storage Requirements 2°C to 8°C
Target ERK1/2
Test Time 1 hrs 20 mins
Total Assay Time 24 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.