Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Immunostain Enhancer

Catalog No. PI46644 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI46644 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI46644 Supplier Thermo Scientific™ Supplier No. 46644
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Increase signal intensity 3- to 12-fold and antibody dilution 5- to 20-fold beyond normal using both chromogenic or fluorescent IHC substrates.

Thermo Scientific™ Pierce Immunostain Enhancer enables significant antibody dilution (5- to 20-fold beyond normal) to increase signal intensity 3- to 12-fold in immunostaining applications with chromogenic or fluorescent substrates.

This ready-to-use enhancer for immunohistochemical staining does not add steps to the workflow because it is used to dilute the primary and secondary antibodies. The Pierce Immunostain Enhancer is compatible with fluorescence and chromogenic detection and routinely increases both signal intensity and detection sensitivity. Signal enhancement is antibody-dependent and typically ranges from 3- to 12-fold. Because of the strong signal enhancement, the Pierce Immunostain Enhancer reduces the amount of antibody required to achieve optimal detection.

Highlights:

  • Save precious antibody – Pierce Immunostain Enhancer allows the customer to use only a fraction of antibody to achieve the same signal as with conventional immunodetection

  • Convenience – simply replace your current antibody dilution buffer with Pierce Immunostain Enhancer (unlike other signal enhancement methods which require additional steps)

  • Increased signal intensity and sensitivity – provides 3- to 12- fold increase in signal intensity and sensitivity for improved visualization of the antigen of interest in cells and tissues

  • Improved specificity – significantly improves signal-to-noise ratio for poor quality and low affinity antibodies

  • Compatible – can be used with chromogenic and fluorescent detection methods

TRUSTED_SUSTAINABILITY

Specifications

Type Immunostain Enhancer
Product Line Pierce
For Use With (Application) Immunohistochemistry
Format Liquid
Reagent Type Signal Enhancers
Content And Storage Upon receipt store at 4°C.
Quantity 20 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.