Learn More
Thermo Scientific™ Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)
Description
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven PRTC HeavyPeptide AQUA Ultimate peptides in five isotopologue versions for the generation of seven standard curves to assess sensitivity and dynamic range of LC-MS/MS systems.
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mix) contains 7 of 15 peptides from the Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88320). Each of the seven peptides has five versions (amino acids labeled with 0, 1, 2, 3, or 4 heavy isotopes) at five distinct concentrations. The 7 x 5 mix can be used with Skyline software to assess dynamic range, linearity, and lower limit of quantitation (LLOQ).
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) is:
• High quality—contains five sets of seven HeavyPeptide AQUA Ultimate peptides, each distinguishable through unique isotopologue labeling
• Optimized—enables scheduling of MS acquisition windows for improved quantification and increased multiplexing
• Quantitative—assesses dynamic range of instrument and instrument sensitivity
• Predictive—enables assessment of chromatography and MS instrument performance
Applications:
• Optimization of LC & MS parameters
• Standard for data-dependent acquisition (DDA)
• Standard for targeted MS (e.g., parallel-reaction monitoring (PRM)
• Identification of total peptide elution window
• Assessment dynamic range of nano or capillary flow LC-MS/MS systems
Specifications
Specifications
| Description | Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture) (5 pmol/μL) |
| Product Line | Pierce |
| Concentration | 5 pmol/μL |
| Type | LC-MS/MS System Standard |
| Quantity | 100 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.