Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PDPH (3-(2-pyridyldithio)propionyl hydrazide)

Catalog No. PI22301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22301 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22301 Supplier Thermo Scientific™ Supplier No. 22301
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crosslink sulfhydryls to periodate-treated glycoprotein sugars via cleavable disulfide bonds using this short crosslinker containing pyridyldithiol and hydrazide groups.

Thermo Scientific™ Pierce PDPH is a short, pyridyldithiol-and-hydrazide crosslinker for reversibly conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).

Highlights:

  • Pyridyldisulfide group provides a cleavable disulfide bond
  • Oxidized carbohydrate-reactive hydrazide
  • Reactive groups: pyridyldisulfide and hydrazide
  • Reactive toward: sulfhydryl groups and carbohydrate
  • Use with EDC for reactivity toward carboxyl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Carbohydrate-Sulfhydryl
Cleavable By Thiols
Description PDPH
Molecular Weight (g/mol) 229.32
PEGylated No
Spacer Arm Length 9.2 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Hydrazide, Pyridyldisulfide
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.