Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein Concentrators PES, 50K MWCO, 0.5–100 mL

Catalog No. PI88541
Change view
Click to view available options
Quantity:
10 Concentrators
24 Concentrators
25 Concentrators
4 Concentrators
Sample Volume:
≤0.5 mL
2 to 6 mL
20 to 100 mL
5 to 20 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sample Volume
PI88541 24 Concentrators 5 to 20 mL
PI88504 25 Concentrators ≤0.5 mL
PI88538 10 Concentrators 2 to 6 mL
PI88539 24 Concentrators 2 to 6 mL
PI88540 10 Concentrators 5 to 20 mL
PI88542 4 Concentrators 20 to 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. PI88541 Supplier Thermo Scientific™ Supplier No. 88541
Add to Cart
Edge
Add to Cart

Thermo Scientific Pierce Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of proteins over 50 KDa, in sample volumes from 0.5 to 100 mL.

Easily and quickly concentrate 50K molecular weight cutoff (MWCO) protein and biological samples with Thermo Scientific Pierce Protein Concentrators PES (polyethersulfone), which are disposable centrifugation devices that enable fast sample recovery and a short processing time. These concentrator columns can be used with standard microcentrifuge equipment, and samples can be collected without the need for invert spinning. Samples can be concentrated with or without desalting.

For protein and other biological samples, such as nucleic acids with a 50K MWCO, Thermo Scientific Pierce Protein Concentrators PES offer rapid protein concentration, excellent sample recovery, and instrument compatibility. These centrifugal devices feature a high-flux polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples. This product has a molecular-weight cutoff of 50K and can be used for processing sample volumes of 0.5–100 mL.

Features of 50K MWCO Protein Concentrators PES include:
Rapid protein concentration—sample concentration of 10- to 30-fold can be achieved in 15 or fewer minutes
Excellent protein recovery—typical protein recovery is >90% (when the sample mass is at least two-fold greater than the posted MWCO)
Instrument-compatibility—a standard microcentrifuge with swinging (except 88504) or fixed-angle rotors can be used; sample can be collected and recovered without invert spinning
Versatile—easily perform sample concentration, diafiltration, or buffer-exchange of biological samples

For easy identification, the MWCO value is etched on the side of each device. To assist with sample addition and recovery, a clear window with graduations is located on the side of each device enabling visual determination of the retentate volume. The vertical design of the membrane allows high-flux rates, low non-specific binding, and negligible protein aggregation on the membrane and provides reliable and consistent results for processing sample volumes between 0.5 and 100 mL. Typical protein recovery is 90% when the mass of the proteins in the sample is at least two-fold greater than the MWCO of the device used. For example, when using IgG (MW ∼150K) samples, use the 5K, 10K, 30K, or 50K MWCO Concentrators. Sample concentration of 10- to 30-fold can be achieved within 15 minutes.

Protein Concentrators PES applications
• Protein concentration of tissue culture media, antiserum, or monoclonal antibody preparations
• Concentration of protein peaks following gel-permeation chromatography
• Removal of unincorporated label following protein modification reactions
• Concentration and desalt/buffer-exchange after elution of protein from ion-exchange, hydrophobic interaction (HIC), metal chelate, or affinity chromatography columns
• Separation of different sized molecules

Pierce Protein Concentrators PES contain a membrane rated for retaining molecules with a molecular weight at least two-fold greater than the MWCO rating of the PES membrane within the device. Reduced recovery may occur when using a concentrator with molecules smaller than the recommended mass. Protein recovery will vary depending on the specific protein in the sample and its starting concentration.

The dead-stop volume for the 0.5 mL, 2–6 mL, 5–20 mL, and 20–100 mL concentrators is approximately 15 μL, 30 μL, 50 μL, and 350 μL, respectively.

To achieve >90% recovery of protein with the 0.5 mL concentrators, the minimum protein sample concentration should be 0.05 mg/mL. For all other concentrators, the minimum protein sample concentration should be 0.1 mg/mL.

Optimal centrifuges and speeds
The 0.5 mL concentrators can be used effectively at a maximum centrifugal force of 15,000 × g using a fixed angle centrifuge only.

For the 2-6 mL concentrators, swinging-bucket or fixed-angle centrifuge models can be used. Swinging bucket centrifuges capable of generating 2,000–4,000 × g and a maximum centrifugal force of 4,000 × g are recommended. With fixed-angle centrifuges, models capable of generating 5,000–10,000 × g and a maximum centrifugal force of 8,000 × g are recommended.

For the 5-20 mL volume concentrators, swinging-bucket or fixed-angle centrifuge models can be used. Swinging-bucket centrifuges capable of generating 2,000–5,000 × g and a maximum centrifugal force of 4,000 × g are recommended. With fixed-angle centrifuges, models capable of generating 5,000–8,000 × g and a maximum centrifugal force of 6,000 × g are recommended.

For the 20-100 mL concentrators, only swinging-bucket centrifuges capable of generating 1,000–2,000 × g are recommended. Use a maximum centrifugal force of 2,000 × g. These concentrators are not recommended for use in fixed-angle rotors.

Centrifugal force, temperature, sample volume, concentration, and viscosity will all affect filtration rate. Centrifugation time should therefore be optimized for each application.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Filter Type Membrane Filter
Format Conical, Spin Column
Material (Membrane) PES
MWCO 50 kDa
Sample Volume 5 to 20 mL
Max. RCF Swinging-bucket rotor: 5,000 x g, Fixed-angle rotor: 8,000 x g
Product Line Pierce
Quantity 24 Concentrators
Shipping Condition Room Temperature
Type Protein Concentrator
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.