Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Pyridine, LC-MS Grade

Catalog No. FER25104
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FER25104 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FER25104 Supplier Thermo Scientific™ Supplier No. 25104

Perform liquid chromatography (HPLC) and mass spectrometry (LC-MS) applications with this 99.5%-pure pyridine solvent (MW 79.1, CAS 110-86-1) with low UV-absorptivity.

Thermo Scientific Pierce Pyridine (PY) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.

Highlights:

Water-miscible organic solvent for a variety of molecular and protein biology methods

High purity, low water, with no peroxides or aldehydes

Ninhydrin test ensures low amines

UV-absorption test confirms suitability for HPLC detection

Liquid reagent packaged by mass in amber glass bottles

Product Details:

General properties of pyridine Alternative names: Azabenzene

Molecular formula: C5H5N

Molecular weight: 79.1

Density: 0.983g/mL

CAS number : 110-86-1 Pierce pyridine is specially purified and each lot is tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

TRUSTED_SUSTAINABILITY

Specifications

Grade LC-MS
Item Name Pyridine
Quantity 100 g
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Form Liquid
Reagent Type Solvent (Pure)
Product Line Pierce
For Use With HPLC, Mass Spectrometry

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.