Learn More
Description
Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein A/G enable probing and detection of antibodies, especially IgG isotypes, from many different species hosts in Western blotting, ELISA, IHC and other immunoassay protocols.
Highlights:
- Contains four Fc-binding domains from Protein A and two from Protein G
- Provides binding for all antibody species and subclasses recognized by either Protein A or Protein G
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Pierce |
| Form | Powder |
| Protein Subtype | Protein A |
| Target | Antibodies |
| Content And Storage | Upon receipt store at -20°C. Product is shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
