Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein A/G

Catalog No. PI77678 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77678 500 mg
PI21186 5 mg
PI77677 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI77678 Supplier Thermo Scientific™ Supplier No. 77678
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detect antibodies, especially IgG isotypes, from many different species hosts with this purified, alkaline phosphatase- or peroxidase-conjugated recombinant Protein A/G.

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein A/G enable probing and detection of antibodies, especially IgG isotypes, from many different species hosts in Western blotting, ELISA, IHC and other immunoassay protocols.

The extended Fc-binding properties of Protein A/G make it a popular tool in the investigation and purification of immunoglobulins. Protein A/G binds to all human IgG subclasses, IgA, IgE, IgM and to a lesser extent IgD; however, it does not bind mouse IgA, IgM or murine serum albumin. Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins. Individual subclasses of mouse monoclonals are most likely to have stronger affinity to this chimeric protein than to either Protein A or Protein G.

Highlights:

  • Contains four Fc-binding domains from Protein A and two from Protein G
  • Provides binding for all antibody species and subclasses recognized by either Protein A or Protein G
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mg
Product Line Pierce
Form Powder
Protein Subtype Protein A
Target Antibodies
Content And Storage Upon receipt store at -20°C. Product is shipped at ambient temperature.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.