Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein G, Peroxidase Conjugated

Catalog No. PI31499 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31499 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31499 Supplier Thermo Scientific™ Supplier No. 31499
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detect mouse and human antibodies, especially IgG isotypes, in immunoassays with this purified, biotinylated or peroxidase-conjugated recombinant Protein G.

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein G enable probing and detection of mouse and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

Protein G is a bacterial cell wall protein isolated from group G Streptococci. DNA sequencing of native Protein G identifies two IgG-binding domains and sites for albumin and cell surface binding. The albumin and cell surface binding domains have been eliminated from Recombinant Protein G to reduce nonspecific binding and, therefore, can be used to separate IgG from crude samples. Optimal binding occurs at pH 5, although binding is also effective at pH 7.0 to 7.2.

Highlights:

  • Contains two Fc-binding domains per protein
  • Better than Protein A for mouse (incl. IgG1), human (incl. IgG3) and rat, goat and cow antibodies
  • Poorer than Protein A for guinea pig, pig, dog and cat
  • Does not bind human IgM, IgD or IgA
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein G, Peroxidase Conjugate
Conjugate HRP
Quantity 0.5 mg
Product Line Pierce
Form Powder
Protein Subtype Protein G
Target Antibodies
Content And Storage Upon receipt store lyophilized product at -20°C. Product is shipped at ambient temperature.
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.