Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fish Serum Blocking Buffer

Catalog No. PI37527
Change view
Click to view available options
Quantity:
500 mL
3 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI37527 500 mL
PI37527X3 3 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI37527 Supplier Thermo Scientific™ Supplier No. 37527
Only null left
Add to Cart
Add to Cart

Thermo Scientific Fish Serum Blocking Buffer is steelhead salmon serum in PBS that is especially useful as a blocking agent in immunohistochemistry (IHC) and other detection methods involving mammalian samples.

Thermo Scientific Fish Serum Blocking Buffer is steelhead salmon serum in PBS that is especially useful as a blocking agent in immunohistochemistry (IHC) and other detection methods involving mammalian samples. Fish Serum Blocking Buffer is effective at stabilizing solutions and samples used for antibody-binding interactions, while at the same time minimizing the possibility of cross-reaction with mammalian sample components.

Features of Fish Serum Blocking Buffer include:
Non-mammalian—fish proteins are less likely to have specific binding interactions with antibodies and other mammalian proteins present in typical methods
Convenient—filtered and stabilized in PBS for compatibility with most assay systems
Easy to use—can be used as supplied or diluted up to 10-fold as needed
Flexible - may be used for many different applications, including as a diluent for antibodies

Fish serum offers several advantages over typical blocking buffers. Because salmon is phylogenetically distant from mammals, which are the source of the antibodies and samples used in most experiments, its serum proteins are less likely to have specific binding interactions with proteins used in the experiments. Fish Serum Blocking Buffer is effective at stabilizing solutions and samples used for antibody-binding interactions while at the same time minimizing the possibility of cross-reaction with mammalian sample components. Try it when use of more-standard sera results in background staining in IHC. In addition, this blocker is effective in various cellular imaging protocols based on visible and near-infrared fluorescence detection. It has been used to decrease background and increase signal-to-noise ratios with near-IR fluorescent probes.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fish Serum Blocking Buffer
Concentration 1X
For Use With (Application) ELISA
Physical Form Liquid
Quantity 500 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.