Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SPB (succinimidyl-[4-(psoralen-8-yloxy)]-butyrate)

Catalog No. PI23013 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI23013 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI23013 Supplier Thermo Scientific™ Supplier No. 23013
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Crosslink amines to DNA with this crosslinker composed of NHS-ester and psoralen reactive groups; conjugates to DNA via light-activated intercalation next to pyrimidine bases.

Thermo Scientific™ Pierce SPB is an NHS-ester and psoralen crosslinker that effectively conjugates primary amines to DNA via light-activated intercalation next to pyrimidine bases.

Highlights:

  • Melting point: 177-178°C
  • Reactive groups: NHS ester and psoralen (Amax= 282nm)
  • Reactive toward: amino groups pyrimidines in DNA or RNA
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Nonselective
Cleavable No
Description SPB
Molecular Weight (g/mol) 385.32
PEGylated No
Spacer Arm Length 8.6 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Not determined
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Psoralen, NHS Ester
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.