Learn More
Thermo Scientific™ Pierce™ Streptavidin, Maleimide-Activated
Description
Thermo Scientific™ Pierce Streptavidin and Conjugates include purified recombinant streptavidin biotin-binding protein, plus standard SA-HRP, SA-AP and fluorescent conjugates for biotin-based assays.
Highlights:
Recombinant streptavidin – high-quality protein, 53kDa, pI equals 6.8 to 7.5
Pure protein and enzyme conjugates – choose pure, unconjugated protein for biotin blocking and controls; enzyme conjugates for substrate-based detection
Fluorophore conjugates – standard fluorescent forms (fluorescein, rhodamine, etc.) for traditional immunofluorescence techniques
Activated preparations – choose a maleimide-activated or hydrazide-activated form to prepare a custom streptavidin conjugate with sulfhydryl- or carbohydrate-containing molecule
Specifications
Specifications
| Conjugate | Streptavidin |
| Description | Pierce Streptavidin, Maleimide Activated |
| Formulation | Lyophilized powder from protein in buffered saline solution |
| Quantity | 1 mg |
| Content And Storage | Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature. |
| Chemical Reactivity | thiol |
| Reactive Moiety | Maleimide |
| Solubility | Soluble in Water |
| Form | Solid |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.