Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format

Catalog No. p-4531027
Change view
Click to view available options
Quantity:
50 mg
10 x 2 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA39268 10 x 2 mg
PI22322 50 mg
PI22122 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIA39268 Supplier Thermo Scientific™ Supplier No. A39268
Only null left
Add to Cart
Edge
Add to Cart

Crosslink amines to sulfhydryls with this mid-length, water-soluble crosslinker made of NHS-ester and maleimide groups at ends of a cyclohexane-containing, 8-atom spacer arm.

Thermo Scientific™ Pierce Sulfo-SMCC is a very popular, water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 Ås).

  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Water-soluble (compare to SMCC), so crosslinking can be done in physiologic solutions
  • High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
  • Cyclohexane bridge confers added stability to the maleimide group making Sulfo-SMCC the ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Description Sulfo-SMCC
Molecular Weight (g/mol) 436.37
PEGylated No
Spacer Arm Length 8.3 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Ambient
Product Line No-Weigh
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short (<10 Å)
Form Solid
Quantity 10 x 2 mg
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.