Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
PIEZO2 Polyclonal Antibody for IHC (P), ICC/IF
Component of mechanosensitive channel required for rapidly adapting mechanically activated (MA) currents. Requires for Merkel-cell mechanotransduction.
Specifications
Specifications
| Antigen | PIEZO2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PIEZO2 |
| Gene Accession No. | Q8CD54, Q9H5I5 |
| Gene Alias | 5930434P17; 9030411M15Rik; 9430028L06Rik; C18orf30; C18orf58; DA3; DA5; Fam38b; FAM38B2; family with sequence similarity 38, member A pseudogene; family with sequence similarity 38, member B; family with sequence similarity 38, member B2; HsT748; HsT771; LOW QUALITY PROTEIN: piezo-type mechanosensitive ion channel component 2; MWKS; piezo type mechanosensitive ion channel component 2; Piezo2; piezo-type mechanosensitive ion channel component 1 pseudogene; piezo-type mechanosensitive ion channel component 2; piezo-type mechanosensitive ion channel component 2 pseudogene; Protein FAM38B; protein PIEZO2 |
| Gene Symbols | PIEZO2 |
| Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
