Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIEZO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572976 Supplier Invitrogen™ Supplier No. PA572976
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIEZO2 Polyclonal Antibody for IHC (P), ICC/IF

Component of mechanosensitive channel required for rapidly adapting mechanically activated (MA) currents. Requires for Merkel-cell mechanotransduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIEZO2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PIEZO2
Gene Accession No. Q8CD54, Q9H5I5
Gene Alias 5930434P17; 9030411M15Rik; 9430028L06Rik; C18orf30; C18orf58; DA3; DA5; Fam38b; FAM38B2; family with sequence similarity 38, member A pseudogene; family with sequence similarity 38, member B; family with sequence similarity 38, member B2; HsT748; HsT771; LOW QUALITY PROTEIN: piezo-type mechanosensitive ion channel component 2; MWKS; piezo type mechanosensitive ion channel component 2; Piezo2; piezo-type mechanosensitive ion channel component 1 pseudogene; piezo-type mechanosensitive ion channel component 2; piezo-type mechanosensitive ion channel component 2 pseudogene; Protein FAM38B; protein PIEZO2
Gene Symbols PIEZO2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1600-1650).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100734606, 307380, 63895, 667742
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.