Learn More
Specifications
Specifications
| Gene ID (Entrez) | 80119 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-97931. For further blocking peptide related protocol, click here . |
| Gene Alias | C15orf20, PIF, PIF1 5-to-3 Prime DNA helicase homolog, RRM3 DNA helicase-like protein |
| Gene Symbol | PIF1 |
| Quantity | 0.1 mg |
| Research Category | Cell Cycle and Replication |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
