Learn More
Enzo Life Sciences Pifithrin-alpha (cyclic). hydrobromide (50mg). CAS: 60477-34-1
SDP
Supplier:  Enzo Life Sciences ALX270287M050
Encompass_Preferred
Stable analog of pifithrin-α (BML-GR325), with biological properties similar to its parent compound, but with reduced cytotoxicity. Inhibitor of p53. Reversibly inhibits p53-dependent transactivation of p53-responsive genes and reversibly blocks p53-mediated apoptosis. Potent STAT6 transcriptional inhibitor. Increases apoptosis in target cells when used in combination with antimicrotubule agents. Formula: C16H16N2S . HBr. MW: 268.4 . 80.9. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in DMSO (100mg/ml). Long Term Storage: -20°C. Handling: Protect from light and moisture. Keep under inert gas.
Catalog No. 50-200-8467
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.