Learn More
Enzo Life Sciences Pifithrin-alpha (5mg). CAS: 63208-82-2
SDP
Supplier:  Enzo Life Sciences BMLGR3250005
Encompass_Preferred
Chemical inhibitor of p53 that protects mice from the side effects of cancer therapy. Reversibly blocks p53-dependent transcriptional activation and apoptosis. Protects against neuronal death in models of stroke and neurodegenerative disorders. Suppresses self-renewal of embryonic stem cells. Alternative name: 1-(4-Methylphenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzothiazolyl)ethanone . HBr, PFT-alpha. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0978
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.