Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pifithrin-mu 98.0+%, TCI America™
SDP

Catalog No. P204810MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P204810MG 10 mg
P2048100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P204810MG Supplier TCI America Supplier No. P204810MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64984-31-2
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD00181531
InChI Key ZZUZYEMRHCMVTB-UHFFFAOYSA-N
Synonym pifithrin-mu, pifithrin-, i, 2-phenylacetylenesulfonamide, phenylethynesulfonamide, phenylacetylenylsulfonamide, 2-phenylethyne-1-sulfonamide, pifithrin mu, pftmu, pifithrin-micro, pifithrin-u
PubChem CID 327653
IUPAC Name 2-phenylethynesulfonamide
SMILES C1=CC=C(C=C1)C#CS(=O)(=O)N

Specifications

Melting Point 137°C
Color White
Quantity 10 mg
Formula Weight 181.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Pifithrin-mu
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.