Learn More
New Pig PIG™ Aerosol Single-Can Recycler
SDP
Catalog No. 18389102 Shop All New Pig Corporation Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
18-389-102 Gray
1 options
This item is not returnable. View return policy
Catalog No. 18-389-102 Supplier New Pig Supplier No. DRM1266
Add to Cart
Add to Cart
This item is not returnable. View return policy
Puncture your aerosol cans for easy, legal recycling to avoid the hassle, regulation and expense of hazardous waste handling.
  • Recycling system easily punctures aerosol cans, rendering them exempt from hazardous waste regulations when properly emptied
  • Eliminates the risks associated with illegal aerosol disposal and the expense of hazardous waste handling
  • Minimizes worker injuries caused by fire, explosion or VOC release from improperly emptied or unemptied cans
  • System works with domed mini, standard and jumbo aerosol cans
  • Combination Coalescing/Carbon Filter captures odors and potentially harmful VOCs released by punctured aerosol cans
  • Timestrip indicator eliminates the guesswork of carbon cartridge changeouts
  • Combination Coalescing/Carbon Filter life is 3 months Angled hardened steel pin punctures cans cleanly without leaving jagged edges
  • Unit mounts directly into 2 in. bung on a 30- or 55-gallon closed-head steel drum; residual liquids drain directly into drum Included ground wire helps prevent static electricity buildup
  • Selling scrap steel can help you meet waste minimization goals at your facility
Specifications
Color Gray
Disposable Reusable
Latex Free Yes
Sterility Non-sterile
Autoclavable Not Autoclavable