Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pig anti-Guinea Pig IgG (H+L) Secondary Antibody, R&D Systems™
SDP

Catalog No. NBP269392 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-69392 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-69392 Supplier R&D Systems Supplier No. NBP269392
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Pig Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Guinea Pig samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgG (H+L)
Applications Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Formulation Lyophilized from PBS (pH 7.2) with No Preservative
Host Species Pig
Immunogen Purified normal IgG isolated from pooled guinea pig serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is directed to the Fc and Fab subunits of the IgG molecule. It includes a certain degree of reactivity with other immunoglobulins via the common Fab portion. It does not react with any non-Ig protein in guinea pig serum, as tested by immunoelectrophoresis and double radial immunodiffusion.
Reconstitution Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.