Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pig anti-Human IgA/IgG/IgM (H+L) Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP269258 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-69258 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-69258 Supplier R&D Systems Supplier No. NBP269258

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Pig Polyclonal Antibody

IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Human samples. It is validated for Immunoelectrophoresis.

Specifications

Antigen IgA/IgG/IgM (H+L)
Applications Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Radioimmunodiffusion, Immunoelectrophoresis
Formulation Lyophilized from PBS (pH 7.2) with No Preservative
Host Species Pig
Immunogen Highly purified IgG and pools of homogenous IgA and IgM isolated from human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is directed to the major isotypes of the human immunoglobulin system (classes and both light chain types) including antibodies to common determinants, to class and to the surface determinants of the common Fab portion, as tested by immunoelectrophoresis and double radial immunodiffusion against pooled serum and purified immunoglobulins. In immunoelectrophoresis and double radial immunodiffusion using various antiserum concentrations against normal human plasma and serum, the characteristic IgG, IgA and IgM precipitin lines are obtained.
Reconstitution Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.