Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pig anti-Human IgG Fc Secondary Antibody, R&D Systems™
SDP

Catalog No. NBP269484 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-69484 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-69484 Supplier R&D Systems Supplier No. NBP269484

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Pig Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Human samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen IgG Fc
Applications Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Formulation Lyophilized from PBS (pH 7.2) with No Preservative
Host Species Pig
Immunogen Purified normal IgG isolated from pooled human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is directed to the Fc subunit of the IgG molecule. In immunoelectrophoresis and double radial immunodiffusion, using various antiserum concentrations against normal human plasma and serum, a characteristic precipitin line is obtained which shows a reaction of identity with the precipitin line obtained with purified IgG. No reaction is obtained with purified IgA, IgM and IgG/Fab fragments or any other serum protein.
Reconstitution Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.