Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pig anti-Human IgM Fc Secondary Antibody, Novus Biologicals™
SDP

Catalog No. p-7084087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-69708 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-69708 Supplier Novus Biologicals Supplier No. NBP269708
Only null left
Add to Cart
Add to Cart

Pig Polyclonal Antibody

IgM Fc Polyclonal antibody specifically detects IgM Fc in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen IgM Fc
Applications Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 1:10 - 1:500
Formulation Lyophilized from PBS (pH 7.2) with No Preservative
Host Species Pig
Immunogen Purified homogenous IgM isolated from human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is directed to the Fc subunit of the IgM molecule which expresses strict isotypic (class) specificity. It does not react with IgG, IgA and IgG/Fab or any non-Ig protein in human serum, as tested by immunoelectrophoresis and double radial immunodiffusion.
Reconstitution Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.