Learn More
Biomatik Corporation Pig Apolipoprotein A2 (APOA2) ELISA Kit
Supplier:  Biomatik Corporation EKU02489
Encompass_Preferred
Pig Apolipoprotein A2 (APOA2) ELISA Kit for detecting APOA2 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 4.69-300ug/ml. Sensitivity: 1.8ug/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1S1A9. Target Synonyms: Apo-A2; Apolipoprotein A-II; ProapoA-II; Truncated apolipoprotein A-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5663
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.