Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig PIG™ Blood, Vomit & Urine Cleanup Kit
SDP

Catalog No. 19041538 Shop All New Pig Corporation Products
Change view
Click to view available options
Capacity (English):
37.4 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-041-538 37.4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-041-538 Supplier New Pig Supplier No. KIT5004
  • $59.80
    $53.65 / Each of 1

    Save $6.15

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blood, Vomit & Urine Cleanup Kit helps you stay clean, safe and sanitary while you take care of bodily fluid spills.

  • Compact design lets you store a spill kit virtually anywhere you need one for fast response
  • Includes absorbent mat pads and loose absorbent to clean up blood, vomit, urine and other bodily fluids (absorbs up to 37 ounces)
  • Biohazard disposal bag helps you control germs and reduce the risk of infection
  • Disposable gloves and mini dustpan also included to aid cleanup
  • Single-use kit is faster and more efficient than dragging a large spill kit to a small cleanup; safer and more effective than using rags or paper towels

Includes:

  • 3 - 10 in. W x 13 in. L PIG™ Absorbent Mat Pad
  • 1 - 3 oz. PIG™ Sheen Clean™ Loose Absorbent
  • 1 - 9.25 x 13 x 4 in. Bag Single Use - Blood, Vomit & Urine Cleanup Kit
  • 1 - 12 in. L SHOWA NITRI-DEX™ Nitrile Gloves (GLV970-XL)
  • 1 - Biohazard Waste Bag 1 - Disposable Scoop and Scraper

Specifications

Description Perlite
Capacity (English) 37.4 oz.
Capacity (Metric) 1.1 L
Material Polypropylene
For Use With (Application) Cleanup Bodily Fluids
Length (English) 9.25 in.
Length (Metric) 23.5 cm
Width (English) 4 in.
Width (Metric) 10.16 cm
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.