Learn More
New Pig PIG Caution Industrial Absorbent Adhesive-Backed Grippy Floor Mat
Shop All New Pig Corporation Products

Description
- Provides traction and safe footing in slip prone areas
- Bright yellow color and caution message draw attention to potential slip hazards from leaks and drips around machinery, equipment, workstations and production areas, even in poorly lit areas
- Adhesive-backed mat stays exactly where you put it with no shifting, bunching up or flipping over to prevent slips, trips and falls
- Sticks to most commercial floor surfaces like cleaned and sealed concrete, vinyl tile, linoleum, ceramic tile and quarry tile
- Tough, spunbond top layer with heavy-duty zigzag stitching stands up to foot and light-wheeled traffic; keep mat in service until it's saturated or shows signs of wear
- Makes it easy to sweep up debris like metal shavings and wood chips while also absorbing common industrial fluids
- Multiple layers of fine-fiber polypropylene safely absorb leaks, drips and overspray of oils, coolants, solvents and water to keep them from being tracked around
- Liquidproof barrier layer prevents absorbed liquid from soaking through to the floor for easier change outs and floor protection
- Easy to cut and install: just roll it out, cut to size, stick it down and apply pressure to smooth it out
- Align seams end to end or side by side to cover large areas; do not overlap seams to avoid premature lifting
- Proprietary adhesive sticks to the floor with a super-tight grip, but peels up easily for quick changeouts; reduces cleanup time to make your facility more productive
- Meets NFPA33 – Static Dissipative and OSHA
- Standards at conditions of 50% RH or higher
- Tested and Certified by the National Floor Safety Institute (NFSI): when used as part of your floor safety program, high-traction surfaces such as PIG Caution Grippy Mat have been proven to reduce slip-and-fall claims by as much as 90%
Specifications
Specifications
| Certifications/Compliance | NFSI Certified, ANSI B101.1- 2009 |
| Color | Yellow |
| Length (English) | 100 ft. |
| Length (Metric) | 30 m |
| Material | Polypropylene |
| Type | Adhesive Floor Mat |
| Width (English) | 32 in. |
| Width (Metric) | 81 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.