Learn More
American Research Products Inc Pig free thyroXine FT4 ELISA
Supplier:  American Research Products Inc CSBEQF027512PI
Encompass
Pig free thyroXine (FT4) ELISA kit from CUSABIO Detection Range 4 pmol/L-64 pmol/L Sensitivity 1 08 pmol/L Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-28886
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.