Learn More
New Pig PIG™ HazMat Spill Kit in 20-Gallon High-Visibility Container
SDP
Catalog No. 18389120 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: wheeled container
Catalog No.
18-389-120 Type: wheeled container
1 options
This item is not returnable. View return policy
Catalog No. 18-389-120 Supplier New Pig Supplier No. KIT312
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-visibility container is easily seen - even in low light or from a distance - to help make spill response even faster.
  • Prepacked containers are organized for easy access
  • Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Lime green drum is easy to locate
  • Lightweight, polypropylene drum resists chemicals and keeps contents clean and dry
  • Bold lettering for quick identification
  • Threaded lid secures contents while providing easy access (not rated as a shipping container)
  • Tamperproof seal helps prevent pilfering of spill response supplies