Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig Lite-Dri™ Loose Absorbent
SDP

Catalog No. 189992540 Shop All New Pig Corporation Products
Change view
Click to view available options
Packaging Quantity:
22 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
18-999-2540 22 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-2540 Supplier New Pig Supplier No. PLP201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use silica-free material to reduce labor and disposal costs

  • Silica-free composition is non-abrasive and does not pose the health hazards associated with breathing in clay dust
  • Non-abrasive composition won't harm finished floors or damage expensive machinery
  • Light-weight absorbent is easier than clay to carry and use without injury
  • Absorbs and retains oils, coolants, solvents and water

Specifications

Capacity (English) 8 gal.
Capacity (Metric) 30.28 L
Material Cellulose
Type Loose Absorbent
Format Powder
For Use With (Application) For most common industrial liquids, is easier to lift and lighter to carry than clay
Packaging Quantity 22 lb.
Length (English) 29 in.
Length (Metric) 73.66 cm
Width (English) 6 in.
Width (Metric) 15.24 cm
Weight (English) 23 lb.
Weight (Metric) 10 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.