Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig PIG™ Mercury Cleanup Wipers
SDP

Catalog No. 18366459 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W):
9 x 8.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-366-459 9 x 8.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-459 Supplier New Pig Supplier No. WIP1404
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pop-up wipers help suppress hazardous vapors.

  • Citrus-based formula is designed for final wipe-downs after a mercury spill has been cleaned up. It helps suppress any remaining vapors to limit worker exposure
  • Effectively removes dangerous residue from hands, tools and hard surfaces that have been exposed to mercury
  • Durable plastic container with snap-tight lid neatly dispenses wipers individually while keeping the remainder clean and moist
  • Note: Avoid contact with metallic nitrates, cyanides, sulfides and strong oxidizers; contact with sodium or calcium hypochlorite creates chlorine gas

Specifications

Color White
Material Polypropylene and Cellulose Blend
Type Mercury Cleaning Kit
For Use With (Application) Cleanup Mercury Residue
For Use With (Equipment) Mercury
Packaging Quantity 10 Pk.
Dimensions (L x W) 9 x 8.5 in.
Weight (English) 0.45 lb.
Weight (Metric) 204 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.