Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig Multi-Purpose Repair Putty
SDP

Catalog No. 19003415 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Epoxy Resin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-003-415 Epoxy Resin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-003-415 Supplier New Pig Supplier No. PTY201
  • $266.00
    $137.65 / Pack of 12

    Save $128.35

    48% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Top-quality resins bond better and stay stronger so repairs last

  • Made with high-quality epoxy resins to bond better and stay stronger; adheres to steel, glass, masonry and most plastics
  • Convenient, premeasured epoxy has Parts A and B in one tube ready for kneading
  • Sets in 20 minutes and cures concrete-hard in one hour for drilling, tapping, sanding or painting
  • Nontoxic, VOC- and solvent-free putty can be used on potable water lines
  • Three- to five-minute working time

Specifications

Color Gray
Material Epoxy Resin
Type Multipurpose Putty
For Use With (Application) Better and stay stronger so repairs last, multipurpose
Length (English) 7 in.
Length (Metric) 17.78 cm
Weight (English) 4 lb.
Weight (Metric) 1.8 kg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.