Learn More
New Pig PIG Oil-Only UV-Resistant Absorbent Barrel Top Mat
SDP
Catalog No. 17987525 Shop All New Pig Corporation Products
Change view
Click to view available options
Weight Capacity:
Heavyweight
Catalog No. Weight Capacity
17-987-525 Heavyweight
1 options
Catalog No. 17-987-525 Supplier New Pig Supplier No. MAT4109
Add to Cart
Add to Cart
Shaped to fit on drum tops to keep them clean, these oil-only absorbent mats withstand direct sun for up to a year without disintegrating.
  • Heavyweight construction is highly durable and absorbent to handle frequent pump drips and spills of oils
  • Soak up pump drips from barrel tops before they become puddles
  • Won't rip, tear or fray when saturated and is UV resistant up to 12 months; provides long-term outdoor use without degradation
  • Box is wall-mount capable for convenient dispensing and storage (template printed on back of box, mounting hardware not included)
  • Precut for perfect fit on 55-gallon steel drums with holes for .75 in. and 2 in. bungs
  • Absorbs and retains oils – not water
  • Wringable and incinerable to help reduce waste
Specifications
Absorbancy 7 gal.
Color White
Material Polypropylene
Type Barrel Top Absorbent Mat
Disposable Single-use
Weight Capacity Heavyweight
Weight (English) 7 lb.
Weight (Metric) 3.2 kg