Learn More
Biomatik Corporation Pig Osteoprotegerin (OPG) ELISA Kit
Supplier:  Biomatik Corporation EKU06437
Encompass_Preferred
Pig Osteoprotegerin (OPG) ELISA Kit for detecting OPG in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A5HMU5. Target Synonyms: TNFRSF11B; TNFRSF11-B; OCIF; OPG; TR1; Tumor Necrosis Factor Receptor Superfamily Member 11b; Osteoclastogenesis Inhibitory Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5903
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.